accessibility menu, dialog, popup

UserName

OHF

CAS RN 371-41-5

IUPAC Name: 4-fluorophenol
Synonyms: 4-fluorophenol P-fluorophenol ccris 665
Molecular Weight (g/mol): 112.1
Molecular Formula: C6H5FO
InChi Key: RHMPLDJJXGPMEX-UHFFFAOYSA-N
SMILES: OC1=CC=C(F)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)