missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenol, 99%

Product Code. 10723671 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10723671 25 g Glass bottle 25g
10519610 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10723671 Supplier Thermo Scientific Acros Supplier No. 119560250
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 371-41-5 | C6H5FO | 112.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 371-41-5
Molecular Formula C6H5FO
Molecular Weight (g/mol) 112.10
MDL Number MFCD00002316
InChI Key RHMPLDJJXGPMEX-UHFFFAOYSA-N
Synonym p-fluorophenol, phenol, 4-fluoro, phenol, p-fluoro, para-fluorophenol, 4-fluoro phenol, 4-fluoro-phenol, ccris 665, 4-fluoranylphenol, fpn
PubChem CID 9732
IUPAC Name 4-fluorophenol
SMILES OC1=CC=C(F)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 44.0°C to 49.0°C
Color Beige-Yellow to Pink
Boiling Point 185.0°C
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4OH
Quantity 25 g
Beilstein 06, 183
Solubility Information Solubility in water: 80g/L (20°C). Other solubilities: soluble in methylene dichloride
Formula Weight 112.1
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 4-Fluorophenol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluorophenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.