accessibility menu, dialog, popup

UserName

NF

CAS RN 372-47-4

IUPAC Name: 3-fluoropyridine
Synonyms: 3-fluoropyridine
Molecular Weight (g/mol): 97.09
Molecular Formula: C5H4FN
InChi Key: CELKOWQJPVJKIL-UHFFFAOYSA-N
SMILES: FC1=CN=CC=C1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (4)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)