missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoropyridine, 99%

Product Code. 10625262 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10625262 1 g Glass bottle 1g
10066393 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10625262 Supplier Thermo Scientific Acros Supplier No. 188300010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 372-47-4 | C5H4FN | 97.092 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372-47-4
MDL Number MFCD00006374
InChI Key CELKOWQJPVJKIL-UHFFFAOYSA-N
Synonym pyridine, 3-fluoro, 3-fluorpyridin, ccris 1717, 3-fluropyridine, 3-fluoro-pyridine, 3-fluoro-pyridinium, pubchem1095, acmc-1coty, 3-fluoropyridine, ksc173o1l
PubChem CID 67794
IUPAC Name 3-fluoropyridine
SMILES C1=CC(=CN=C1)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 1.1300g/mL
Boiling Point 107.0°C to 108.0°C
Flash Point 13°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C5H4FN
Refractive Index 1.4720 to 1.4740
Quantity 1 g
Beilstein 20, IV, 2492
Specific Gravity 1.13
Molecular Weight (g/mol) 97.09
Formula Weight 97.09
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Fluoropyridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Fluoropyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.