accessibility menu, dialog, popup

UserName

H2NN

CAS RN 3731-52-0

IUPAC Name: 1-(pyridin-3-yl)methanamine
Synonyms: 3-pyridinemethaneamine
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: HDOUGSFASVGDCS-UHFFFAOYSA-N
SMILES: NCC1=CC=CN=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)