missing translation for 'onlineSavingsMsg'
Learn More

3-(Aminomethyl)pyridine, 98+%

Product Code. 11460028
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460028 100 g 100g
11450028 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11460028 Supplier Thermo Scientific Alfa Aesar Supplier No. B23634.22
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3731-52-0 | C6H8N2 | 108.144 g/mol

It finds its application as a rubefacient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application as a rubefacient.

Solubility
Fully miscible in water. Soluble in chloroform, ethyl acetate and methanol.

Notes
Air sensitive. Store away from air, carbon dioxide and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3731-52-0
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006412
InChI Key HDOUGSFASVGDCS-UHFFFAOYSA-N
Synonym 3-aminomethyl pyridine, 3-pyridinemethanamine, 3-picolylamine, picolamine, 3-pyridylmethylamine, 3-pyridinylmethanamine, 3-pyridinemethylamine, 3-aminomethylpyridine, pyridin-3-yl methanamine, 3-pyridylmethanamine
PubChem CID 31018
IUPAC Name pyridin-3-ylmethanamine
SMILES C1=CC(=CN=C1)CN

Specifications

Melting Point -21°C
Density 1.062
pH 11 to 12
Boiling Point 226°C
Flash Point 100°C (212°F)
Odor Amine-like
Refractive Index 1.551
Quantity 100 g
UN Number UN2735
Beilstein 108056
Sensitivity Air Sensitive
Solubility Information Fully miscible in water. Soluble in chloroform,ethyl acetate and methanol.
Formula Weight 108.14
Percent Purity ≥99%
Chemical Name or Material 3-(Aminomethyl)pyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.