accessibility menu, dialog, popup

UserName

OH(R)O(R)(S)ONON(R)OH

CAS RN 3736-77-4

IUPAC Name: (2R,4R,5R,6S)-5-hydroxy-4-(hydroxymethyl)-3,7-dioxa-1,9-diazatricyclo[6.4.0.0²,⁶]dodeca-8,11-dien-10-one
Synonyms: cyclouridine O2,2'-cyclouridine ANU 2,2'-O-cyclouridine 2,2'-anhydrouridine
Molecular Weight (g/mol): 226.19
Molecular Formula: C9H10N2O5
InChi Key: UUGITDASWNOAGG-CCXZUQQUSA-N
SMILES: OC[C@H]1O[C@@H]2[C@@H](OC3=NC(=O)C=CN23)[C@@H]1O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results