accessibility menu, dialog, popup

UserName

NN

CAS RN 3759-28-2

IUPAC Name: 2-(cyanomethyl)benzonitrile
Synonyms: o-tolunitrile, α-cyano-
Molecular Weight (g/mol): 142.16
Molecular Formula: C9H6N2
InChi Key: GKHSEDFDYXZGCG-UHFFFAOYSA-N
SMILES: N#CCC1=CC=CC=C1C#N
Grade 
  • (3)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)