missing translation for 'onlineSavingsMsg'
Learn More

Homophthalonitrile, tech. 80%

Product Code. 11388785
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388785 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11388785 Supplier Thermo Scientific Alfa Aesar Supplier No. L12419.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3759-28-2 | C9H6N2 | 142.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3759-28-2
Molecular Formula C9H6N2
Molecular Weight (g/mol) 142.16
MDL Number MFCD00001895
InChI Key GKHSEDFDYXZGCG-UHFFFAOYSA-N
Synonym 2-cyanomethyl benzonitrile, homophthalonitrile, 2-cyanobenzyl cyanide, 2-cyanophenylacetonitrile, alpha-cyano-o-tolunitrile, o-cyanobenzyl cyanide, benzeneacetonitrile, 2-cyano, 2-cyanobenzeneacetonitrile, o-cyanophenyl acetonitrile, o-tolunitrile, alpha-cyano
PubChem CID 77368
IUPAC Name 2-(cyanomethyl)benzonitrile
SMILES N#CCC1=CC=CC=C1C#N

Specifications

Melting Point 73°C to 79°C
Quantity 5 g
UN Number UN3439
Beilstein 1940497
Formula Weight 142.16
Percent Purity 90%
Grade Technical
Chemical Name or Material Homophthalonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.