accessibility menu, dialog, popup

UserName

CH3OPSOCH3OH3CSCH3O

CAS RN 3761-41-9

IUPAC Name: O-4-methanesulfinyl-3-methylphenyl O,O-dimethyl phosphorothioate
Synonyms: fenthion sulfoxide
Molecular Weight (g/mol): 294.32
Molecular Formula: C10H15O4PS2
InChi Key: DLAPIMGBBDILHJ-UHFFFAOYNA-N
SMILES: COP(=S)(OC)OC1=CC=C(C(C)=C1)S(C)=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Concentration 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)