accessibility menu, dialog, popup

UserName

CAS RN 380430-70-6

BOOOH3CCH3H3CHOOH

CAS RN 380430-70-6

IUPAC Name: (4-{[(tert-butoxy)carbonyl]oxy}phenyl)boronic acid
Synonyms: 4-[(tert-butoxycarbonyl)oxy]phenylboronic acid
Molecular Weight (g/mol): 238.05
Molecular Formula: C11H15BO5
InChi Key: HZOYMUMPDHVQOH-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)OC1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results