missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-(tert-Butoxycarboxy)benzeneboronic acid, 97%

Product Code. 15419825
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15419825 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15419825 Supplier Alfa Aesar™ Supplier No. H52469.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 380430-70-6
Molecular Formula C11H15BO5
Molecular Weight (g/mol) 238.05
MDL Number MFCD03095144
InChI Key HZOYMUMPDHVQOH-UHFFFAOYSA-N
Synonym 4-tert-butoxycarbonyl oxy phenyl boronic acid, 4-tert-butoxycarboxyphenylboronic acid, 4-tert-butoxycarbonyl oxy phenylboronic acid, pubchem1761, 4-tert-butoxycarboxyphenyl boronic acid, 4-2-methylpropan-2-yl oxycarbonyloxy phenyl boronic acid, 4-tert-butoxycarbonyloxyphenyl boronic acid, 4-tert-butoxycarbonyl oxy phenyl boronicacid
PubChem CID 2773306
SMILES CC(C)(C)OC(=O)OC1=CC=C(C=C1)B(O)O

Specifications

Melting Point 140°C to 146°C
Quantity 250 mg
IUPAC Name [4-[(2-methylpropan-2-yl)oxycarbonyloxy]phenyl]boronic acid
Formula Weight 238.05
Percent Purity 97%
Chemical Name or Material 4-(tert-Butoxycarboxy)benzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.