CAS RN 38377-38-7
CAS RN 38377-38-7
IUPAC Name:
4-fluorophenyl carbonochloridate
Synonyms:
4-fluorophenyl carbonochloridate
Molecular Weight (g/mol):
174.56
Molecular Formula:
C7H4ClFO2
InChi Key:
MSBGPEACXKBQSX-UHFFFAOYSA-N
SMILES:
FC1=CC=C(OC(Cl)=O)C=C1
1
–
2
of
2
results
1
4-Fluorophenyl chloroformate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Fluorophenyl chloroformate, 98%, Thermo Scientific Chemicals
CAS: 38377-38-7 Molecular Formula: C7H4ClFO2 Molecular Weight (g/mol): 174.56 MDL Number: MFCD00013257 InChI Key: MSBGPEACXKBQSX-UHFFFAOYSA-N Synonym: 4-fluorophenyl chloroformate,4-fluorophenyl carbonochloridate,acmc-1ad3b,4-fluorophenyl chlorocarbonate,4-fluoro phenyl chloroformate,chloroformic acid p-fluorophenyl ester,carbonochloridic acid 4-fluorophenyl ester PubChem CID: 2733256 IUPAC Name: (4-fluorophenyl) carbonochloridate SMILES: C1=CC(=CC=C1OC(=O)Cl)F