missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenyl chloroformate, 98%, Thermo Scientific Chemicals

Product Code. 10414473 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10414473 1 g Glass bottle 1g
10093620 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10414473 Supplier Thermo Scientific Acros Supplier No. 308320010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38377-38-7
Molecular Formula C7H4ClFO2
Molecular Weight (g/mol) 174.56
MDL Number MFCD00013257
InChI Key MSBGPEACXKBQSX-UHFFFAOYSA-N
Synonym 4-fluorophenyl chloroformate, 4-fluorophenyl carbonochloridate, acmc-1ad3b, 4-fluorophenyl chlorocarbonate, 4-fluoro phenyl chloroformate, chloroformic acid p-fluorophenyl ester, carbonochloridic acid 4-fluorophenyl ester
PubChem CID 2733256
IUPAC Name (4-fluorophenyl) carbonochloridate
SMILES C1=CC(=CC=C1OC(=O)Cl)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.3290g/mL
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula ClCO2C6H4F
Refractive Index 1.4923 to 1.4943
Quantity 1 g
Specific Gravity 1.329
Formula Weight 174.56
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Fluorophenyl chloroformate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.