CAS RN 3843-74-1
CAS RN 3843-74-1
IUPAC Name:
methyl (2E)-3-(3,4-dihydroxyphenyl)prop-2-enoate
Synonyms:
methyl caffeate
Molecular Weight (g/mol):
194.19
Molecular Formula:
C10H10O4
InChi Key:
OCNYGKNIVPVPPX-HWKANZROSA-N
SMILES:
COC(=O)\C=C\C1=CC=C(O)C(O)=C1
1
–
1
of
1
results
1
Methyl caffeate
CAS: 3843-74-1 Molecular Formula: C10H10O4 Molecular Weight (g/mol): 194.186 MDL Number: MFCD00210468 InChI Key: OCNYGKNIVPVPPX-HWKANZROSA-N Synonym: methyl caffeate,methyl 3-3,4-dihydroxyphenyl acrylate,caffeic acid methyl ester,e-methyl 3-3,4-dihydroxyphenyl acrylate,methylcaffeate,methyl 3,4-dihydroxycinnamate,caffeic acid, methyl ester,unii-n79173b9hf,methyl 2e-3-3,4-dihydroxyphenyl prop-2-enoate,chembl17001 PubChem CID: 689075 ChEBI: CHEBI:6856 IUPAC Name: methyl (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate SMILES: COC(=O)C=CC1=CC(=C(C=C1)O)O