missing translation for 'onlineSavingsMsg'
Learn More

Methyl caffeate

Product Code. 15485949
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15485949

Brand: Thermo Scientific Alfa Aesar J63786.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibitor of protein tyrosine kinase | CAS: 3843-74-1 | C10H10O4 | 194.186 g/mol

Inhibitor of protein tyrosine kinase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3843-74-1
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00210468
InChI Key OCNYGKNIVPVPPX-HWKANZROSA-N
Synonym methyl caffeate, methyl 3-3,4-dihydroxyphenyl acrylate, caffeic acid methyl ester, e-methyl 3-3,4-dihydroxyphenyl acrylate, methylcaffeate, methyl 3,4-dihydroxycinnamate, caffeic acid, methyl ester, unii-n79173b9hf, methyl 2e-3-3,4-dihydroxyphenyl prop-2-enoate, chembl17001
PubChem CID 689075
ChEBI CHEBI:6856
IUPAC Name methyl (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate
SMILES COC(=O)C=CC1=CC(=C(C=C1)O)O

Specifications

Melting Point 158°C to 161°C
Color Yellow
Quantity 250 mg
Merck Index 14,1635
Formula Weight 194.19
Physical Form Powder
Chemical Name or Material Methyl caffeate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.