accessibility menu, dialog, popup

UserName

ONOClOO

CAS RN 38464-20-9

IUPAC Name: 4-chloro-3-nitro-2H-chromen-2-one
Synonyms: 4-chloro-3-nitrochromen-2-one
Molecular Weight (g/mol): 225.58
Molecular Formula: C9H4ClNO4
InChi Key: OFLRQEKOAGDHKT-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(Cl)C2=CC=CC=C2OC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results