missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-nitrocoumarin, 98%

Product Code. 11332097
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332097 5 g 5g
11322097 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332097 Supplier Thermo Scientific Alfa Aesar Supplier No. L07908.06
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 38464-20-9 | Molecular Formula: C9H4ClNO4 | Molecular Weight: 225.584 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Chloro-3-nitrocoumarin, is used in synthesis and antimicrobial activity of new 4-heteroarylamino coumarin derivatives that contain nitrogen and sulfur as heteroatoms.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38464-20-9
Molecular Formula C9H4ClNO4
Molecular Weight (g/mol) 225.584
MDL Number MFCD00051670
InChI Key OFLRQEKOAGDHKT-UHFFFAOYSA-N
Synonym 4-chloro-3-nitrocoumarin, 4-chloro-3-nitro-2h-chromen-2-one, 4-chloro-3-nitro-chromen-2-one, 3-nitro-4-chlorocoumarin, pubchem8672, acmc-1ahgg, 4-chloranyl-3-nitro-chromen-2-one, 4-chloro-3-nitro-1-benzopyran-2-one, 2h-1-benzopyran-2-one,4-chloro-3-nitro
PubChem CID 688979
IUPAC Name 4-chloro-3-nitrochromen-2-one
SMILES C1=CC=C2C(=C1)C(=C(C(=O)O2)[N+](=O)[O-])Cl

Specifications

Melting Point ∽162°C (decomposition)
Quantity 5 g
Beilstein 883649
Solubility Information Insoluble in water.
Formula Weight 225.59
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-nitrocoumarin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.