accessibility menu, dialog, popup

UserName

CAS RN 385-02-4

FOOHNOO

CAS RN 385-02-4

IUPAC Name: 2-fluoro-6-nitrobenzoic acid
Synonyms: 2-fluoro-6-nitrobenzoic acid
Molecular Weight (g/mol): 185.11
Molecular Formula: C7H4FNO4
InChi Key: MPDZCNPDHUUPRL-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(C=CC=C1F)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results