missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-nitrobenzoic acid, 98%

Product Code. 15449448
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15449448 100 g 100g
15429448 5 g 5g
15439448 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15449448 Supplier Thermo Scientific Alfa Aesar Supplier No. H63013.22
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 385-02-4 | C7H4FNO4 | 185.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 385-02-4
Molecular Formula C7H4FNO4
Molecular Weight (g/mol) 185.11
MDL Number MFCD01862079
InChI Key MPDZCNPDHUUPRL-UHFFFAOYSA-N
Synonym 2-fluoro-6-nitrobenzoic acid, 2-fluoro-6-nitrobenzoicacid, 6-fluoro-2-nitrobenzoic acid, 2-carboxy-3-fluoronitrobenzene, 2-fluoro-6-nitro-benzoic acid, benzoic acid, 2-fluoro-6-nitro, pubchem1308, 6-fluoro-nitrobenzoic acid, acmc-209j0j, ksc222c6b
PubChem CID 603758
SMILES C1=CC(=C(C(=C1)F)C(=O)O)[N+](=O)[O-]

Specifications

Quantity 100 g
Formula Weight 185.11
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Fluoro-6-nitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.