accessibility menu, dialog, popup

UserName

FFFSClOOFFF

CAS RN 39234-86-1

IUPAC Name: 3,5-bis(trifluoromethyl)benzene-1-sulfonyl chloride
Synonyms: 3,5-bis(trifluoromethyl)benzenesulfonyl chloride
Molecular Weight (g/mol): 312.61
Molecular Formula: C8H3ClF6O2S
InChi Key: BTRCVKADYDVSLI-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC(=C1)C(F)(F)F)S(Cl)(=O)=O
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results