missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di(trifluoromethyl)benzene-1-sulfonyl chloride, 97%, Maybridge

Product Code. 10572652
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10572652 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10572652 Supplier Thermo Scientific Maybridge Supplier No. JP00937DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 39234-86-1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H3ClF6O2S
Linear Formula (CF3)2C6H3SO2Cl
MDL Number MFCD00014725
Quantity 1 g
Synonym 3,5-bis trifluoromethyl benzenesulfonyl chloride, 3,5-bis trifluoromethyl benzene-1-sulfonyl chloride, 3,5-bis trifluoromethyl benzenesulphonyl chloride, 3,5-di trifluoromethyl benzene-1-sulfonyl chloride, 3,5-ditrifluoromethylbenzenesulfonyl chloride, benzenesulfonyl chloride, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenylsulfonyl chloride, 3,5-bis-trifluoromethyl-benzenesulfonyl chloride, 3,5-di trifluoromethyl benzene sulfonyl chloride, pubchem2724
InChI Key BTRCVKADYDVSLI-UHFFFAOYSA-N
SMILES C1=C(C=C(C=C1C(F)(F)F)S(=O)(=O)Cl)C(F)(F)F
IUPAC Name 3,5-bis(trifluoromethyl)benzenesulfonyl chloride
Molecular Weight (g/mol) 312.61
PubChem CID 520945
Formula Weight 312.62
Percent Purity 97%
Chemical Name or Material 3, 5-Di(trifluoromethyl)benzene-1-sulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.