accessibility menu, dialog, popup

UserName

ONOClNOOFFF

CAS RN 393-75-9

IUPAC Name: 2-chloro-1,3-dinitro-5-(trifluoromethyl)benzene
Synonyms: 1,3-DINO2 2-CL 5-CF3 benzene
Molecular Weight (g/mol): 270.55
Molecular Formula: C7H2ClF3N2O4
InChi Key: HFHAVERNVFNSHL-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC(=C1Cl)[N+]([O-])=O)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results