missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3,5-dinitrobenzotrifluoride, TRC

Product Code. 30138239
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
Unit Size:
100g
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138239 250 g 250g
30135999 100 g 100g
30008798 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138239 Supplier Toronto Research Chemicals Supplier No. TRCC367130250G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 393-75-9
Molecular Formula C7 H2 Cl F3 N2 O4
Molecular Weight (g/mol) 270.55
Synonym 2-Chloro-1,3-dinitro-5-(trifluoromethyl)-benzene, 4-Chloro-α,α,α-trifluoro-3,5-dinitro-toluene, 1,3-Dinitro-2-chloro-5-(trifluoromethyl)benzene, 1-Chloro-2,6-dinitro-4-(trifluoromethyl)benzene, 2,6-Dinitro-4-(trifluoromethyl)chlorobenzene, 2-Chloro-1,3-dinitro-5-(trifluoromethyl)benzene, 3,5-Dinitro-4-chlorobenzotrifluoride, 3,5-Dinitro-4-chlorotrifluoromethylbenzene, 4-(Trifluoromethyl)-2,6-dinitrochlorobenzene, 4-Chloro-3,5-dinitro-α,α,α-trifluorotoluene, 4-Chloro-3,5-dinitrobenzotrifluoride, 4-Chloro-α,α,α-trifluoro-3,5-dinitrotoluene, Chloralin, NSC 88274
IUPAC Name 2-chloro-1,3-dinitro-5-(trifluoromethyl)benzene
SMILES [O-][N+](=O)c1cc(cc(c1Cl)[N+](=O)[O-])C(F)(F)F

Specifications

Quantity 250 g
InChI Formula InChI=1S/C7H2ClF3N2O4/c8-6-4(12(14)15)1-3(7(9,10)11)2-5(6)13(16)17/h1-2H
Formula Weight 269.9655
Chemical Name or Material 4-Chloro-3,5-dinitrobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.