accessibility menu, dialog, popup

UserName

CAS RN 3943-77-9

H3COOOCH3OH3C

CAS RN 3943-77-9

IUPAC Name: ethyl 3,4-dimethoxybenzoate
Synonyms: ethyl 3,4-dimethoxybenzoate
Molecular Weight (g/mol): 210.23
Molecular Formula: C11H14O4
InChi Key: AYYNUGSDPRDVCH-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(OC)C(OC)=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results