missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,4-dimethoxybenzoate, 98+%

Product Code. 11357675
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357675 100 g 100g
11347675 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357675 Supplier Thermo Scientific Alfa Aesar Supplier No. L02355.22
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3943-77-9 | C11H14O4 | 210.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3943-77-9
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.23
MDL Number MFCD00015150
InChI Key AYYNUGSDPRDVCH-UHFFFAOYSA-N
Synonym benzoic acid, 3,4-dimethoxy-, ethyl ester, ethyl veratrate, 3,4-dimethoxybenzoic acid ethyl ester, veratric acid, ethyl ester 7ci,8ci, veratric acid, ethyl ester, veratrumsaureathylester, acmc-1cme0, ethyl-3,4-dimethoxybenzoate, 4-10-00-01469 beilstein handbook reference, ksc581g5p
PubChem CID 77546
IUPAC Name ethyl 3,4-dimethoxybenzoate
SMILES CCOC(=O)C1=CC=C(OC)C(OC)=C1

Specifications

Melting Point 42°C to 45°C
Quantity 100 g
Beilstein 3118897
Formula Weight 210.23
Percent Purity ≥98%
Chemical Name or Material Ethyl 3,4-dimethoxybenzoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.