CAS RN 395-82-4
IUPAC Name:
4-fluoro-2-methoxybenzoic acid
Synonyms:
4-fluoro-2-methoxybenzoic acid
Molecular Weight (g/mol):
170.14
Molecular Formula:
C8H7FO3
InChi Key:
UUQDNAPKWPKHMK-UHFFFAOYSA-N
SMILES:
COC1=CC(F)=CC=C1C(O)=O
1
–
1
of
1
results
1
4-Fluoro-2-methoxybenzoic acid, 99%
CAS: 395-82-4 Molecular Formula: C8H7FO3 Molecular Weight (g/mol): 170.14 MDL Number: MFCD00673010 InChI Key: UUQDNAPKWPKHMK-UHFFFAOYSA-N Synonym: 2-methoxy-4-fluorobenzoic acid,4-fluoro-2-methoxy-benzoic acid,4-fluoro-o-anisic acid,benzoic acid, 4-fluoro-2-methoxy,pubchem4969,acmc-209j6b,ksc494s1h,4-fluoro-2-methoxybenzoicacid,rarechem al be 1023,4-fluoro-2-methoxy benzoic acid PubChem CID: 2774545 IUPAC Name: 4-fluoro-2-methoxybenzoic acid SMILES: COC1=CC(F)=CC=C1C(O)=O