CAS RN 3968-92-1
CAS RN 3968-92-1
IUPAC Name:
(butan-2-yl)triphenylphosphanium bromide
Synonyms:
triphenyl(sec-butyl)phosphanium bromide
Molecular Weight (g/mol):
399.31
Molecular Formula:
C22H24BrP
InChi Key:
HKWAYXKPYJOYEL-UHFFFAOYNA-M
SMILES:
[Br-].CCC(C)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1