missing translation for 'onlineSavingsMsg'
Learn More

(2-Butyl)triphenylphosphonium bromide, 96%

Product Code. 15418799
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 15418799

Brand: Thermo Scientific Alfa Aesar L01532.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3968-92-1 | C22H24BrP | 399.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3968-92-1
Melting Point 227°C to 237°C
Molecular Formula C22H24BrP
MDL Number MFCD00050236
Quantity 5 g
Beilstein 3580097
Sensitivity Hygroscopic
Synonym 2-butyl triphenylphosphonium bromide, sec-butyltriphenylphosphonium bromide, triphenyl sec-butyl phosphanium bromide, acmc-1cswd, butan-2-yl triphenyl phosphanium bromide, phosphonium, 1-methylpropyl triphenyl-, bromide 1:1
InChI Key HKWAYXKPYJOYEL-UHFFFAOYNA-M
SMILES [Br-].CCC(C)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name butan-2-yl(triphenyl)phosphanium;bromide
Molecular Weight (g/mol) 399.31
PubChem CID 3083826
Formula Weight 399.32
Percent Purity 96%
Chemical Name or Material (2-Butyl)triphenylphosphonium bromide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.