CAS RN 39925-19-4
CAS RN 39925-19-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1-beta-D-Ribofuranosyl-1H-1,2,4-triazole-3-carboxylic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Ribavirin Carboxylic Acid, TRC
CAS: 39925-19-4 Molecular Formula: C8 H11 N3 O6 Molecular Weight (g/mol): 245.19 Synonym: 1-beta-D-Ribofuranosyl-1H-1,2,4-triazole-3-carboxylic Acid,Ribavirin Imp. A (EP) IUPAC Name: 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,4-triazole-3-carboxylic acid SMILES: OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc(n2)C(=O)O