accessibility menu, dialog, popup

UserName

H2NOBr

CAS RN 4001-73-4

IUPAC Name: 2-bromobenzamide
Synonyms: 2-bromobenzamide
Molecular Weight (g/mol): 200.04
Molecular Formula: C7H6BrNO
InChi Key: NHNAEZDWNCRWRW-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CC=C1Br
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)