missing translation for 'onlineSavingsMsg'
Learn More

2-Bromobenzamide, 97%, Maybridge

Product Code. 10473393
Change view
Click to view available options
Amber Glass Bottle; 10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10473393 Amber Glass Bottle; 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10473393 Supplier Thermo Scientific Maybridge Supplier No. RDP00161EA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4001-73-4
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C7H6BrNO
Linear Formula BrC6H4C(=O)NH2
Quantity 10 g
Synonym benzamide, 2-bromo, o-bromobenzamide, benzamide, o-bromo, 2-bromo-benzamide, 2-bromobenzamide, bromobenzamide, pubchem3733, maybridge1_005867, acmc-209j9x
InChI Key NHNAEZDWNCRWRW-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)C(=O)N)Br
IUPAC Name 2-bromobenzamide
Molecular Weight (g/mol) 200.035
PubChem CID 77616
Formula Weight 200.03
Percent Purity 97%
Chemical Name or Material 2-Bromobenzamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.