accessibility menu, dialog, popup

UserName

H3COOOCH3OH3C

CAS RN 4038-15-7

IUPAC Name: (2,4-dimethoxyphenyl)(4-methoxyphenyl)methanone
Synonyms: (2,4-dimethoxyphenyl)(4-methoxyphenyl)methanone
Molecular Weight (g/mol): 272.3
Molecular Formula: C16H16O4
InChi Key: VFTDVICZBGDMMB-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(=O)C1=CC=C(OC)C=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results