missing translation for 'onlineSavingsMsg'
Learn More

2,4,4'-Trimethoxybenzophenone, 99%, Thermo Scientific™

Product Code. 11323157
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11323157 50 g 50g
11313157 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11323157 Supplier Thermo Scientific Alfa Aesar Supplier No. L13580.18
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4038-15-7
Molecular Formula C16H16O4
Molecular Weight (g/mol) 272.3
MDL Number MFCD00100089
InChI Key VFTDVICZBGDMMB-UHFFFAOYSA-N
Synonym 2,4,4'-trimethoxybenzophenone, 2,4-dimethoxyphenyl 4-methoxyphenyl methanone, acmc-20aofo, maybridge1_005563, 2,4-dimethoxy-4'-methoxybenzophenone, 2, 4-dimethoxy-4'-methoxybenzophenone, 2,4-dimethoxyphenyl 4-methoxyphenyl ketone, 2,4-dimethoxyphenyl-4-methoxyphenyl methanone, methanone, 2,4-dimethoxyphenyl 4-methoxyphenyl, 2,4-dimethoxy-phenyl-4-methoxy-phenyl-methanone
PubChem CID 254174
IUPAC Name (2,4-dimethoxyphenyl)-(4-methoxyphenyl)methanone
SMILES COC1=CC=C(C=C1)C(=O)C2=C(C=C(C=C2)OC)OC

Specifications

Melting Point 71°C to 73°C
Quantity 50 g
Beilstein 1991889
Formula Weight 272.3
Percent Purity 99%
Chemical Name or Material 2,4,4'-Trimethoxybenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.