CAS RN 405165-61-9
CAS RN 405165-61-9
IUPAC Name:
7-(3-aminoazepan-1-yl)-8-chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride
Synonyms:
besifloxacin hydrochloride
Molecular Weight (g/mol):
430.3
Molecular Formula:
C19H22Cl2FN3O3
InChi Key:
PMQBICKXAAKXAY-UHFFFAOYNA-N
SMILES:
Cl.NC1CCCCN(C1)C1=C(F)C=C2C(=O)C(=CN(C3CC3)C2=C1Cl)C(O)=O
1
–
2
of
2
results
1
Thermo Scientific Chemicals Besifloxacin hydrochloride
CAS: 405165-61-9 Molecular Formula: C19H22Cl2FN3O3
2
Besifloxacin Hydrochloride, MedChemExpress
MedChemExpress Besifloxacin hydrochloride is a fourth-generation fluoroquinolone antibiotic.