accessibility menu, dialog, popup

UserName

CH3NNH2S

CAS RN 4104-75-0

IUPAC Name: 1-methyl-1-phenylthiourea
Synonyms: urea,1-methyl-1-phenyl-2-thio
Molecular Weight (g/mol): 166.24
Molecular Formula: C8H10N2S
InChi Key: MCWZNJNWGQPUGL-UHFFFAOYSA-N
SMILES: CN(C(N)=S)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results