missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-phenylthiourea, 97%, Thermo Scientific™

Product Code. 11465756
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Unit Size:
10g
2.5g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465756 50 g 50g
11445756 2.5 g 2.5g
11455756 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11465756 Supplier Thermo Scientific Alfa Aesar Supplier No. B20824.18
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used to produce S-Ethyl-N-methyl-N-phenyl-isothiourea; hydriodide by heating. In the presence of solvent acetone.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4104-75-0
Molecular Formula C8H10N2S
Molecular Weight (g/mol) 166.242
MDL Number MFCD00086629
InChI Key MCWZNJNWGQPUGL-UHFFFAOYSA-N
Synonym n-methyl-n-phenylthiourea, 1-methyl-1-phenyl-thiourea, thiourea, methylphenyl, urea,1-methyl-1-phenyl-2-thio, amino methylphenylamino methane-1-thione, acmc-1akxy, maybridge1_000090, n-methyl-n-phenyl-thiourea, n-phenyl-n-methyl-thiourea, thiourea, n-methyl-n-phenyl
PubChem CID 723631
IUPAC Name 1-methyl-1-phenylthiourea
SMILES CN(C1=CC=CC=C1)C(=S)N

Specifications

Melting Point 100°C to 104°C
Odor Odorless
Quantity 50 g
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 166.24
Percent Purity 97%
Chemical Name or Material N-Methyl-N-phenylthiourea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.