accessibility menu, dialog, popup

UserName

CH3OO

CAS RN 4133-34-0

IUPAC Name: 7-methoxy-1,2,3,4-tetrahydronaphthalen-2-one
Synonyms: 7-methoxy-3,4-dihydro-1H-naphthalen-2-one
Molecular Weight (g/mol): 176.22
Molecular Formula: C11H12O2
InChi Key: XEAPZXNZOJGVCZ-UHFFFAOYSA-N
SMILES: COC1=CC=C2CCC(=O)CC2=C1
Boiling Point 
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)