missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxy-2-tetralone, 95%, Thermo Scientific™

Product Code. 10617251 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10617251 5 g Glass bottle 5g
10792315 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10617251 Supplier Thermo Scientific Acros Supplier No. 294970050
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 4133-34-0
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.22
MDL Number MFCD00001730
InChI Key XEAPZXNZOJGVCZ-UHFFFAOYSA-N
Synonym 7-methoxy-2-tetralone, 7-methoxy-3,4-dihydronaphthalen-2 1h-one, 3,4-dihydro-7-methoxy-2 1h-naphthalenone, 7-methoxy-3,4-dihydronaphthalen-2-one, 7-methoxy-1,2,3,4-tetrahydronaphthalen-2-one, 7-methoxyl-2-tetralone, 7-methoxytetralin-2-one, 2 1h-naphthalenone, 3,4-dihydro-7-methoxy, 7-methoxy-1,3,4-trihydronaphthalen-2-one, 3,4-dihydro-7-methoxynaphthalen-2 1h-one
PubChem CID 77785
IUPAC Name 7-methoxy-3,4-dihydro-1H-naphthalen-2-one
SMILES COC1=CC2=C(CCC(=O)C2)C=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow to Orange
Density 1.1300g/mL
Boiling Point 124°C to 126°C (1.5 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5590 to 1.563
Quantity 5 g
Specific Gravity 1.13
Formula Weight 176.22
Percent Purity 95%
Physical Form Liquid After Melting
Chemical Name or Material 7-Methoxy-2-tetralone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.