accessibility menu, dialog, popup

UserName

H3COOOOCH3CH3H3C

CAS RN 42726-73-8

IUPAC Name: 1-tert-butyl 3-methyl propanedioate
Synonyms: 1-tert-butyl 3-methyl propanedioate
Molecular Weight (g/mol): 174.2
Molecular Formula: C8H14O4
InChi Key: XPSYZCWYRWHVCC-UHFFFAOYSA-N
SMILES: COC(=O)CC(=O)OC(C)(C)C
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)