missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl methyl malonate, 95%, Thermo Scientific Chemicals

Product Code. 15455898
Change view
Click to view available options
Quantity:
2 mL
10 mL
Unit Size:
10mL
2mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15455898 10 mL 10mL
15445898 2 mL 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455898 Supplier Thermo Scientific Alfa Aesar Supplier No. H59540.AB
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 42726-73-8 | Molecular Formula: C8H14O4 | Molecular Weight: 174.196 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42726-73-8
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.196
MDL Number MFCD00042856
InChI Key XPSYZCWYRWHVCC-UHFFFAOYSA-N
Synonym tert-butyl methyl malonate, t-butyl methyl malonate, 1-tert-butyl 3-methyl propanedioate, acmc-20ajkv, tert-butyl methylmalote, pubchem20476, methyl tert-butyl malonate, ksc493c8h, tert-butyl methyl malonaoate
PubChem CID 2733872
IUPAC Name 3-O-tert-butyl 1-O-methyl propanedioate
SMILES CC(C)(C)OC(=O)CC(=O)OC

Specifications

Density 1.03
Boiling Point 80°C (11 mmHg)
Flash Point 79°C (174°F)
Linear Formula CH3OCOCH2COOC(CH3)3
Refractive Index 1.4155
Quantity 10 mL
Beilstein 1772136
Formula Weight 174.19
Percent Purity 95%
Chemical Name or Material tert-Butyl methyl malonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.