missing translation for 'onlineSavingsMsg'
Learn More

tert.-Butyl methyl malonate, 95%

Product Code. 10783065 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
1mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10783065 25 mL Glass bottle 25mL
10249253 1 mL Glass bottle 1mL
10567011 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10783065 Supplier Thermo Scientific Acros Supplier No. 339390250
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 42726-73-8 | C8H14O4 | 174.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42726-73-8
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.2
MDL Number MFCD00042856
InChI Key XPSYZCWYRWHVCC-UHFFFAOYSA-N
Synonym tert-butyl methyl malonate, t-butyl methyl malonate, 1-tert-butyl 3-methyl propanedioate, acmc-20ajkv, tert-butyl methylmalote, pubchem20476, methyl tert-butyl malonate, ksc493c8h, tert-butyl methyl malonaoate
PubChem CID 2733872
IUPAC Name 3-O-tert-butyl 1-O-methyl propanedioate
SMILES CC(C)(C)OC(=O)CC(=O)OC

Specifications

Melting Point -10°C
Color Colorless to Yellow
Density 1.0300g/mL
Boiling Point 80°C (11.0 mmHg)
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula H3COOCCH2COOC(CH3)3
Refractive Index 1.4140 to 1.417
Quantity 25 mL
Specific Gravity 1.03
Formula Weight 174.2
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material tert.-Butyl methyl malonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.