accessibility menu, dialog, popup

UserName

H3COFNH2

CAS RN 437-83-2

IUPAC Name: 3-fluoro-2-methoxyaniline
Synonyms: 3-fluoro-2-methoxyaniline
Molecular Weight (g/mol): 141.15
Molecular Formula: C7H8FNO
InChi Key: RCYMPYMITUEHOJ-UHFFFAOYSA-N
SMILES: COC1=C(N)C=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results