missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-2-methoxyaniline, 97%, Thermo Scientific Chemicals

Product Code. 15444907
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15444907 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15444907 Supplier Thermo Scientific Alfa Aesar Supplier No. H32214.MD
 Restricted Item
Request Bulk or Custom Format

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 437-83-2
Molecular Formula C7H8FNO
Molecular Weight (g/mol) 141.145
MDL Number MFCD00007759
InChI Key RCYMPYMITUEHOJ-UHFFFAOYSA-N
Synonym 3-fluoro-o-anisidine, 3-fluoro-2-methoxy-phenylamine, 2-amino-6-fluoroanisole, 3-fluoro-2-methoxyphenylamine, benzenamine, 3-fluoro-2-methoxy, 3-fuloro-2-methoxyaniline, 3-fluoro-2-methoxy-benzenamine, 3-fluoro-2-methoxybenzenenamine, pubchem2256, pubchem3021
PubChem CID 581109
IUPAC Name 3-fluoro-2-methoxyaniline
SMILES COC1=C(C=CC=C1F)N

Specifications

Quantity 250 mg
UN Number UN2810
Formula Weight 141.15
Percent Purity 97%
Chemical Name or Material 3-Fluoro-2-methoxyaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.