accessibility menu, dialog, popup

UserName

CAS RN 438050-23-8

CH3OFOHF

CAS RN 438050-23-8

IUPAC Name: (4-ethoxy-2,6-difluorophenyl)methanol
Synonyms: (4-ethoxy-2,6-difluorophenyl)methanol
Molecular Weight (g/mol): 188.17
Molecular Formula: C9H10F2O2
InChi Key: DVBOKVWHRGLKBD-UHFFFAOYSA-N
SMILES: CCOC1=CC(F)=C(CO)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results