missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-2,6-difluorobenzyl alcohol, JRD, 97%, Thermo Scientific™

Product Code. 15465697
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15465697 250 mg 250mg
15455697 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15465697

Brand: Thermo Scientific Alfa Aesar H32920.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 438050-23-8
Molecular Formula C9H10F2O2
Molecular Weight (g/mol) 188.174
MDL Number MFCD09258696
InChI Key DVBOKVWHRGLKBD-UHFFFAOYSA-N
Synonym 4-ethoxy-2,6-difluorophenyl methanol, 4-ethoxy-2,6-difluorobenzyl alcohol, 4-ethoxy-2,6-difluorobenzylalcohol, 4-ethoxy-2,6-difluoro-benzylalcohol, benzenemethanol, 4-ethoxy-2,6-difluoro, 4-ethoxy-2,6-difluorophenyl methan-1-ol, 4-ethoxy-2,6-difluorobenzyl alcohol, jrd
PubChem CID 22219845
IUPAC Name (4-ethoxy-2,6-difluorophenyl)methanol
SMILES CCOC1=CC(=C(C(=C1)F)CO)F

Specifications

Quantity 250 mg
Formula Weight 188.17
Percent Purity 97%
Chemical Name or Material 4-Ethoxy-2,6-difluorobenzyl alcohol, JRD
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.