accessibility menu, dialog, popup

UserName

CH3OHOH2N

CAS RN 4389-45-1

IUPAC Name: 2-amino-3-methylbenzoic acid
Synonyms: 2-amino-3-methylbenzoic acid 3-methylanthranil acid
Molecular Weight (g/mol): 151.17
Molecular Formula: C8H9NO2
InChi Key: WNAJXPYVTFYEST-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C(O)=O)=C1N
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (3)
  • (6)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)