accessibility menu, dialog, popup

UserName

CAS RN 4418-26-2

NaH3COCOOOCH3

CAS RN 4418-26-2

IUPAC Name: sodium 3-acetyl-6-methyl-2,4-dioxo-3,4-dihydro-2H-pyran-3-ide
Synonyms: sodium dehydroacetate
Molecular Weight (g/mol): 190.13
Molecular Formula: C8H7NaO4
InChi Key: YIOCQGHBBNGBND-UHFFFAOYSA-N
SMILES: [Na+].CC(=O)[C-]1C(=O)OC(C)=CC1=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)