missing translation for 'onlineSavingsMsg'
Learn More

Sodium dehydroacetate, 97%

Product Code. 11440157
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11440157

Brand: Thermo Scientific Alfa Aesar B21060.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4418-26-2 | C8H7NaO4

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Sodium dehydroacetate is used in the food industry as a preservative for fruits, vegetables, fruit juice, honey, beer, milk, protein and fat products. Its antimicrobial properties play a vital role in personal care products such as skin care, suntan, sunscreen, shaving, tooth paste, hair and nail care products, and eye and facial makeup. It is used as a plasticizer.

Solubility
Soluble in water, propylene glycol and methanol. Insoluble in acetone and benzene.

Notes
Incompatible strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4418-26-2
Molecular Formula C8H7NaO4
MDL Number MFCD00040583

Specifications

Melting Point ∼295°C (decomposition)
pH 8.8 to 9.3
Quantity 500 g
Synonym 3-Acetyl-4-hydroxy-6-methyl-2-pyrone Sodium Salt; Dehydroacetic Acid Sodium Salt
Solubility Information Soluble in water,propylene glycol and methanol. Insoluble in acetone and benzene.
Formula Weight 190.13
Percent Purity 97%
Chemical Name or Material Sodium Dehydroacetate
compliance-icons
Product Identifier
  • Sodium dehydroacetate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Sodium dehydroacetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.