missing translation for 'onlineSavingsMsg'
Learn More

Dehydroacetic acid sodium salt, 99%, Thermo Scientific Chemicals

Product Code. 10242662 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10242662 500 g Plastic bottle 500g
10494975 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10242662 Supplier Thermo Scientific Acros Supplier No. 364855000
  • ONLINE EXCLUSIVE PRICE
    139.00 €
    129.00 € / 500g
    Save 10.00 €
    7% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4418-26-2 | Molecular Formula: C8H7O4Na | Molecular Weight: 190.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4418-26-2
Molecular Formula C8H7O4Na
MDL Number MFCD00040583
Synonym 3-Acetyl-6-methyl-2H-pyran-2, 4(3H)-dione sodium salt

Specifications

Melting Point 295.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 500 g
Formula Weight 190.13
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Dehydroacetic acid sodium salt, 99%
compliance-icons
Product Identifier
  • Dehydroacetic acid sodium salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.